siRNA Details
| Gene Symbol: Gskip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178613.3 | NP_848728.2 | 2 | 183 | ||
| GenBank mRNA | AK017049.1 | - | 707 | |||
| AK020785.1 | - | 2 | 121 | |||
| AK029484.1 | - | 2 | 95 | |||
| AK042664.1 | - | 2 | 144 | |||
| AK079775.1 | - | 2 | 129 | |||
| AK168962.1 | - | 2 | 147 | |||
| AK171097.1 | - | 2 | 160 | |||
| BC031494.1 | - | 2 | 151 | 3156862 | ||

Gskip
Genomic Map