siRNA Details
| Gene Symbol: Ppcdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176831.4 | NP_789801.1 | 7 | 758 | ||
| GenBank mRNA | AK036042.1 | - | 7 | 741 | ||
| AK039846.1 | - | 2 | 132 | |||
| AK046751.1 | - | 4 | 810 | |||
| AK078825.1 | - | 7 | 959 | |||
| BC004779.1 | - | 7 | 969 | |||
| BC052928.1 | - | 6 | 707 | 5255304 | ||
| BC089351.1 | - | 6 | 722 | 30603115 | ||
| BU053206.1 | - | 4 | 354 | |||

Ppcdc
Genomic Map