siRNA Details
| Gene Symbol: Ppcdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176831.4 | NP_789801.1 | 4 | 371 | ||
| GenBank mRNA | AK036042.1 | - | 4 | 354 | ||
| AK038628.1 | - | 4 | 349 | |||
| AK046751.1 | - | 1 | 205 | |||
| AK078825.1 | - | 4 | 354 | |||
| BC004779.1 | - | 4 | 364 | |||
| BC052928.1 | - | 3 | 320 | 5255304 | ||
| BC089351.1 | - | 3 | 335 | 30603115 | ||

Ppcdc
Genomic Map