siRNA Details
| Gene Symbol: Luc7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025881.3 | NP_080157.1 | 3, 4 | 384 | ||
| NM_028190.3 | NP_082466.2 | 3, 4 | 384 | |||
| GenBank mRNA | AK010539.1 | - | 3, 4 | 336 | ||
| AK017655.1 | - | 3, 4 | 344 | |||
| AK160293.1 | - | 3, 4 | 385 | |||
| AK211801.1 | - | 4, 5 | 312 | |||
| BC019194.1 | - | 4, 5 | 377 | |||
| BC034135.1 | - | 2, 3 | 550 | 5003664 | ||
| BC055875.1 | - | 3, 4 | 1367 | |||
| BQ572015.1 | - | 1, 2 | 36 | |||
| BY726224.1 | - | 4, 5 | 442 | |||

Luc7l
Genomic Map