siRNA Details
| Gene Symbol: Yaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024189.6 | NP_077151.3 | 3 | 378 | ||
| GenBank mRNA | AB100452.1 | - | 3 | 368 | ||
| AB100453.1 | - | 5 | 437 | |||
| AK005008.1 | - | 3 | 362 | |||
| AK011702.1 | - | 4 | 449 | |||
| AK012787.1 | - | 3 | 284 | |||
| AK148634.1 | - | 3 | 322 | |||
| AK179353.1 | - | 1 | 75 | |||
| AK184096.1 | - | 3 | 163 | |||
| BC002192.1 | - | 3 | 251 | 3488691 | ||

Yaf2
Genomic Map