siRNA Details
Gene Symbol: Polr3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164082.1 | NP_001157554.1 | 6 | 1000 | ||
NM_025945.3 | NP_080221.3 | 7 | 1032 | |||
GenBank mRNA | AK013165.1 | - | 6 | 917 | ||
AK156587.1 | - | 7 | 984 | |||
BC016102.1 | - | 7 | 1008 | 4911475 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
