siRNA Details
| Gene Symbol: Polr3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164082.1 | NP_001157554.1 | 5 | 624 | ||
| NM_025945.3 | NP_080221.3 | 6 | 656 | |||
| GenBank mRNA | AK013165.1 | - | 5 | 541 | ||
| AK156587.1 | - | 6 | 608 | |||
| BC016102.1 | - | 6 | 632 | 4911475 | ||
| CA548436.1 | - | 5 | 573 | |||

Polr3d
Genomic Map