siRNA Details
| Gene Symbol: Rnf141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025999.3 | NP_080275.2 | 4 | 419 | ||
| GenBank mRNA | AF353167.1 | - | 4 | 449 | ||
| AK006791.1 | - | 4 | 459 | |||
| AK011839.1 | - | 4 | 408 | |||
| AK015505.1 | - | 4 | 420 | |||
| AK139910.1 | - | 4 | 413 | |||
| AK154417.1 | - | 4 | 443 | |||
| BC018553.1 | - | 4 | 413 | 3582546 | ||

Rnf141
Genomic Map