siRNA Details
| Gene Symbol: Prl8a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023332.3 | NP_075821.1 | 6 | 674 | ||
| GenBank mRNA | AF158744.1 | - | 6 | 659 | ||
| AK005409.1 | - | 6 | 675 | |||
| AK005494.1 | - | 6 | 675 | |||
| BC064762.1 | - | 6 | 642 | 30013931 | ||

Prl8a9
Genomic Map