siRNA Details
| Gene Symbol: Spaca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290443.1 | NP_001277372.1 | 4, 5 | 574 | ||
| NM_026293.3 | NP_080569.1 | 4, 5 | 576 | |||
| GenBank mRNA | AK006187.1 | - | 4, 5 | 577 | ||
| AK007270.1 | - | 4, 5 | 548 | |||
| AK016011.1 | - | 4, 5 | 569 | |||
| BC048388.1 | - | 1, 2 | 233 | |||
| BC147079.1 | - | 4, 5 | 573 | 9056040 | ||
| BC147080.1 | - | 4, 5 | 573 | 9056039 | ||
| BY706285.1 | - | 4, 5 | 575 | |||

Spaca1
Genomic Map