siRNA Details
| Gene Symbol: 4930544G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161773.1 | NP_001155245.1 | 494 | |||
| GenBank mRNA | AK006363.1 | - | 1 | 454 | ||
| AK016035.1 | - | 1 | 456 | |||
| BC107181.1 | - | 1 | 403 | |||
| BC107182.1 | - | 1 | 402 | |||
| EU477372.1 | - | 1 | 282 | |||

4930544G11Rik
Genomic Map