siRNA Details
Gene Symbol: Caap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026368.2 | NP_080644.2 | 6 | 817 | ||
GenBank mRNA | AK076487.1 | - | 6 | 772 | ||
AK140382.1 | - | 6 | 818 | |||
AK150311.1 | - | 7 | 727 | |||
AK160297.1 | - | 6 | 820 | |||
AK160572.1 | - | 6 | 818 | |||
BC020067.1 | - | 6 | 805 | 3991510 |
