siRNA Details
| Gene Symbol: Limd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172397.3 | NP_765985.1 | 5 | 460 | ||
| GenBank mRNA | AK012581.1 | - | 5 | 434 | ||
| AK032430.1 | - | 5 | 461 | |||
| AK040486.1 | - | 5 | 461 | |||
| AK049809.1 | - | 5 | 430 | |||
| AK167349.1 | - | 5 | 430 | |||
| AK169829.1 | - | 5 | 431 | |||
| AK171794.1 | - | 5 | 739 | |||
| AK187911.1 | - | 2 | 160 | |||
| AK188791.1 | - | 2 | 159 | |||
| BC068130.1 | - | 5 | 404 | 6413206 | ||

Limd2
Genomic Map