siRNA Details
| Gene Symbol: Zcchc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175126.4 | NP_780335.1 | 1 | 1374 | ||
| GenBank mRNA | AK053859.1 | - | 1 | 1111 | ||
| BC055755.1 | - | 1 | 1357 | 6412435 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcchc3| Gene Symbol: Zcchc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175126.4 | NP_780335.1 | 1 | 1374 | ||
| GenBank mRNA | AK053859.1 | - | 1 | 1111 | ||
| BC055755.1 | - | 1 | 1357 | 6412435 | ||