siRNA Details
| Gene Symbol: Gcsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026572.3 | NP_080848.1 | 3, 4 | 312 | ||
| GenBank mRNA | AK003189.1 | - | 397 | |||
| AK003788.2 | - | 3, 4 | 309 | |||
| AK009543.1 | - | 1, 2 | 76 | |||
| AK019983.1 | - | 3, 4 | 313 | |||
| AK050373.1 | - | 3, 4 | 325 | |||
| BC014745.1 | - | 3, 4 | 316 | 4239181 | ||
| Gene Symbol: Gm9199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BE853472.1 | - | 1 | 401 | ||

Gcsh
Genomic Map