siRNA Details
| Gene Symbol: Gcsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026572.3 | NP_080848.1 | 5 | 489 | ||
| GenBank mRNA | AK003189.1 | - | 575 | |||
| AK003788.2 | - | 5 | 486 | |||
| AK009543.1 | - | 3 | 253 | |||
| AK019983.1 | - | 5 | 490 | |||
| AK050373.1 | - | 5 | 502 | |||
| AK178705.1 | - | 2 | 113 | |||
| BC014745.1 | - | 5 | 493 | 4239181 | ||

Gcsh
Genomic Map