siRNA Details
| Gene Symbol: Mfsd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026660.2 | NP_080936.1 | 5 | 615 | ||
| GenBank mRNA | AK018736.1 | - | 5 | 628 | ||
| AK089055.1 | - | 5 | 632 | |||
| AK152438.1 | - | 5 | 601 | |||
| BC023012.1 | - | 5 | 615 | 5376230 | ||
| BC064775.1 | - | 5 | 513 | 30023877 | ||

Mfsd10
Genomic Map