siRNA Details
| Gene Symbol: Ccdc163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026714.2 | NP_080990.2 | 5 | 633 | ||
| GenBank mRNA | AI606993.1 | - | 1 | 165 | ||
| AK002768.1 | - | 5 | 535 | |||
| AK016995.1 | - | 4 | 1103 | |||
| BC022718.1 | - | 4 | 607 | |||
| BC022733.1 | - | 5 | 631 | 4506628 | ||
| BG297132.1 | - | 5 | 630 | |||

Ccdc163
Genomic Map