siRNA Details
| Gene Symbol: Ccdc163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026714.2 | NP_080990.2 | 3 | 296 | ||
| GenBank mRNA | AK002768.1 | - | 3 | 198 | ||
| AK016995.1 | - | 2 | 766 | |||
| BC022718.1 | - | 2 | 270 | |||
| BC022733.1 | - | 3 | 294 | 4506628 | ||
| BG297132.1 | - | 3 | 293 | |||
| DT911344.1 | - | 3 | 270 | |||

Ccdc163
Genomic Map