siRNA Details
| Gene Symbol: 1110032A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023483.3 | NP_075972.2 | 3 | 547 | ||
| GenBank mRNA | AJ250230.1 | - | 3 | 497 | ||
| AK004016.1 | - | 3 | 848 | |||
| AK015671.1 | - | 3 | 487 | |||
| AK043513.1 | - | 3 | 548 | |||
| AK079767.1 | - | 2426 | ||||
| AK158460.1 | - | 3 | 536 | |||
| CT010205.1 | - | 3 | 294 | |||

1110032A03Rik
Genomic Map