siRNA Details
| Gene Symbol: 1110032A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023483.3 | NP_075972.2 | 4 | 654 | ||
| GenBank mRNA | AJ250230.1 | - | 4 | 604 | ||
| AK004016.1 | - | 4 | 955 | |||
| AK015671.1 | - | 4 | 594 | |||
| AK043513.1 | - | 4 | 655 | |||
| AK079767.1 | - | 2533 | ||||
| AK158460.1 | - | 4 | 643 | |||
| AK166202.1 | - | 4 | 619 | |||
| BC032012.1 | - | 4 | 628 | |||
| CT010205.1 | - | 4 | 401 | |||

1110032A03Rik
Genomic Map