siRNA Details
Gene Symbol: Mospd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254762.1 | NP_001241691.1 | 3 | 670 | ||
NM_030037.2 | NP_084313.1 | 4 | 823 | |||
GenBank mRNA | AK004495.1 | - | 1 | 68 | ||
AK088731.1 | - | 3 | 661 | |||
BC003880.1 | - | 3 | 680 | 3496491 | ||
BC034167.1 | - | 4 | 808 | |||
BC039620.1 | - | 4 | 804 | 5051528 | ||
BE861793.1 | - | 3 | 297 | |||
BQ445267.1 | - | 2 | 236 | |||
DV656015.1 | - | 1 | 80 |
