siRNA Details
| Gene Symbol: Snx15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026912.1 | NP_081188.1 | 3 | 257 | ||
| GenBank mRNA | BC016091.1 | - | 3 | 257 | 4503873 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx15| Gene Symbol: Snx15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026912.1 | NP_081188.1 | 3 | 257 | ||
| GenBank mRNA | BC016091.1 | - | 3 | 257 | 4503873 | |