siRNA Details
| Gene Symbol: Trip13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027182.2 | NP_081458.1 | 10 | 1185 | ||
| GenBank mRNA | AK010336.1 | - | 10 | 1147 | ||
| AK142463.1 | - | 1158 | ||||
| AK146877.1 | - | 10 | 1186 | |||
| AK151568.1 | - | 10 | 1121 | |||
| AK212754.1 | - | 2 | 223 | |||
| AK215913.1 | - | 2 | 245 | |||
| BC023834.1 | - | 4 | 393 | |||
| BC064315.1 | - | 10 | 1108 | |||
| BC126946.1 | - | 10 | 1084 | 40090292 | ||

Trip13
Genomic Map