siRNA Details
| Gene Symbol: H1fnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027304.2 | NP_081580.2 | 1 | 552 | ||
| GenBank mRNA | AB016273.1 | - | 1 | 524 | ||
| AK006402.1 | - | 1 | 291 | |||
| AY496853.1 | - | 1 | 550 | |||
| BC052363.1 | - | 1 | 538 | 6773647 | ||

H1fnt
Genomic Map