siRNA Details
| Gene Symbol: Fam189a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183087.4 | NP_898910.2 | 10 | 1488 | ||
| GenBank mRNA | AK017751.1 | - | 4 | 594 | ||
| AK138167.1 | - | 8 | 984 | |||
| AK172974.1 | - | 10 | 1260 | |||
| BC049085.1 | - | 7 | 810 | |||
| BC147145.1 | - | 2 | 202 | |||
| BC147146.1 | - | 2 | 202 | |||

Fam189a1
Genomic Map