siRNA Details
| Gene Symbol: 4921504E06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027600.4 | NP_081876.2 | 17 | 1692 | ||
| GenBank mRNA | AK014808.1 | - | 17 | 1693 | ||
| BC132142.1 | - | 17 | 1682 | 40130419 | ||
| BC137968.1 | - | 17 | 1682 | 8860987 | ||

4921504E06Rik
Genomic Map