siRNA Details
| Gene Symbol: Lrrc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027941.1 | NP_082217.1 | 8 | 899 | ||
| GenBank mRNA | AK005720.1 | - | 8 | 951 | ||
| BC100519.1 | - | 8 | 899 | 30919449 | ||
| BC116419.1 | - | 7 | 732 | |||
| BC116420.1 | - | 7 | 739 | 40103399 | ||
| JF934738.1 | - | 8 | 850 | |||

Lrrc34
Genomic Map