siRNA Details
| Gene Symbol: Osgin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027950.1 | NP_082226.1 | 6 | 837 | ||
| GenBank mRNA | AB221676.1 | - | 5 | 664 | ||
| AK005892.1 | - | 4 | 600 | |||
| AK156249.1 | - | 1 | 1326 | |||
| BC006032.1 | - | 5 | 1178 | |||
| BC022135.1 | - | 6 | 837 | 5099665 | ||
| BC091765.1 | - | 1 | 45 | |||
| BC110674.1 | - | 6 | 822 | |||
| BC132440.1 | - | 6 | 806 | 40130717 | ||
| BC138192.1 | - | 6 | 806 | 8861211 | ||

Osgin1
Genomic Map