siRNA Details
| Gene Symbol: Btbd17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028055.4 | NP_082331.1 | 3 | 1112 | ||
| GenBank mRNA | AK005160.1 | - | 3 | 1118 | ||
| AK032100.1 | - | 3 | 1114 | |||
| AK046050.1 | - | 3 | 1113 | |||
| BC057599.1 | - | 2 | 2389 | |||
| BC060053.1 | - | 2 | 2420 | |||
| BC122876.1 | - | 3 | 1095 | 40107633 | ||

Btbd17
Genomic Map