siRNA Details
| Gene Symbol: Shq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012978.1 | - | 10 | 1239 | ||
| BC052657.1 | - | 7 | 898 | 30042589 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001134713, XM_008763162, BC168172.

Shq1
Genomic Map