siRNA Details
| Gene Symbol: Snx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026998.3 | NP_081274.2 | 10 | 881 | ||
| GenBank mRNA | AK008135.1 | - | 5 | 346 | ||
| AK013158.1 | - | 10 | 882 | |||
| AK030764.1 | - | 4 | 211 | |||
| BC061028.1 | - | 11 | 922 | 6594585 | ||

Snx6
Genomic Map