siRNA Details
| Gene Symbol: B3gnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028189.3 | NP_082465.3 | 2 | 647 | ||
| GenBank mRNA | AK008674.1 | - | 686 | |||
| AK144117.1 | - | 637 | ||||
| AK144180.1 | - | 2 | 619 | |||
| AY037785.1 | - | 616 | ||||
| AY039028.1 | - | 1 | 566 | |||
| BC026418.1 | - | 2 | 639 | 4218953 | ||

B3gnt3
Genomic Map