siRNA Details
| Gene Symbol: Brf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028193.3 | NP_082469.2 | 16, 17 | 2090 | ||
| GenBank mRNA | AK010890.1 | - | 3, 4 | 339 | ||
| BC037224.1 | - | 16, 17 | 1873 | 3257364 | ||
| BC058112.1 | - | 6, 7 | 686 | |||

Brf1
Genomic Map