siRNA Details
| Gene Symbol: Brf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028193.3 | NP_082469.2 | 15 | 1888 | ||
| GenBank mRNA | AK010890.1 | - | 2 | 137 | ||
| BC037224.1 | - | 15 | 1671 | 3257364 | ||
| BC058112.1 | - | 5 | 484 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brf1| Gene Symbol: Brf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028193.3 | NP_082469.2 | 15 | 1888 | ||
| GenBank mRNA | AK010890.1 | - | 2 | 137 | ||
| BC037224.1 | - | 15 | 1671 | 3257364 | ||
| BC058112.1 | - | 5 | 484 | |||