siRNA Details
| Gene Symbol: Rhno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011109.1 | - | 2 | 381 | ||
| AK011167.1 | - | 2 | 483 | |||
| AK020031.1 | - | 2 | 538 | |||
| AK029454.1 | - | 2 | 371 | |||
| AK146167.1 | - | 2 | 260 | |||
| AK156717.1 | - | 3 | 803 | |||
| AK168313.1 | - | 732 | ||||
| BC027368.1 | - | 2 | 687 | |||
| BE655013.1 | - | 2 | 289 | |||

Rhno1
Genomic Map