siRNA Details
| Gene Symbol: Rhno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011109.1 | - | 3 | 546 | ||
| AK011167.1 | - | 3 | 647 | |||
| AK029454.1 | - | 3 | 536 | |||
| AK156717.1 | - | 4 | 968 | |||
| AK168313.1 | - | 897 | ||||
| BC005726.1 | - | 1 | 71 | |||
| BC027368.1 | - | 3 | 852 | |||

Rhno1
Genomic Map