siRNA Details
Gene Symbol: Zfp566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152814.2 | NP_690027.1 | 4 | 1064 | ||
GenBank mRNA | AK012367.1 | - | 4 | 1041 | ||
BC019790.1 | - | 4 | 951 | 5151196 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152814.2 | NP_690027.1 | 4 | 1064 | ||
GenBank mRNA | AK012367.1 | - | 4 | 1041 | ||
BC019790.1 | - | 4 | 951 | 5151196 |