siRNA Details
| Gene Symbol: Fam98a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133747.2 | NP_598508.2 | 8 | 1175 | ||
| GenBank mRNA | AK012997.1 | - | 8 | 1109 | ||
| AK046481.1 | - | 7 | 1177 | |||
| AK146215.1 | - | 8 | 1119 | |||
| AK146761.1 | - | 8 | 1141 | |||
| AK146955.1 | - | 8 | 1088 | |||
| AK167224.1 | - | 8 | 1059 | |||
| BC004626.1 | - | 8 | 1072 | 3585698 | ||

Fam98a
Genomic Map