siRNA Details
| Gene Symbol: Hspb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028394.2 | NP_082670.1 | 2 | 132 | ||
| GenBank mRNA | AK013636.1 | - | 2 | 109 | ||
| AK135163.1 | - | 1 | 491 | |||
| AK157651.1 | - | 2 | 133 | |||
| BC021897.1 | - | 2 | 168 | 3710627 | ||
| BC034465.1 | - | 2 | 168 | |||

Hspb11
Genomic Map