siRNA Details
Gene Symbol: Tmem138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007197.1 | - | 2 | 342 | ||
AK013694.1 | - | 1 | 243 | |||
AK031921.1 | - | 2 | 253 | |||
AK050471.1 | - | 2 | 246 | |||
BC058237.1 | - | 2 | 507 | 6307765 | ||
BC107026.2 | - | 1 | 215 | 40050148 | ||
BC132574.1 | - | 1 | 109 | 40130851 | ||
BC132578.1 | - | 1 | 109 | 40130855 | ||
BC145197.1 | - | 1 | 109 |
