siRNA Details
| Gene Symbol: Tmem138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013694.1 | - | 3 | 504 | ||
| AK031921.1 | - | 4 | 441 | |||
| AK050471.1 | - | 4 | 507 | |||
| BC058237.1 | - | 4 | 768 | 6307765 | ||
| BC107026.2 | - | 3 | 476 | 40050148 | ||
| BC132574.1 | - | 3 | 370 | 40130851 | ||
| BC132578.1 | - | 3 | 370 | 40130855 | ||
| BC145197.1 | - | 3 | 297 | |||

Tmem138
Genomic Map