siRNA Details
| Gene Symbol: Pradc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163427.1 | NP_001156899.1 | 2, 3 | 294 | ||
| NM_028505.2 | NP_082781.1 | 2, 3 | 294 | |||
| GenBank mRNA | AK006658.1 | - | 2, 3 | 260 | ||
| BC115452.2 | - | 2, 3 | 240 | 40100110 | ||
| BG088874.2 | - | 2, 3 | 293 | |||
| CJ075544.1 | - | 2, 3 | 318 | |||

Pradc1
Genomic Map