siRNA Details
Gene Symbol: Prox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175198.4 | NP_780407.1 | 5 | 2033 | ||
GenBank mRNA | AK029978.1 | - | 5 | 1990 | ||
AK051700.1 | - | 5 | 2034 | |||
BC119330.1 | - | 4 | 1777 | 8734079 | ||
BC119332.1 | - | 4 | 1777 | 8734081 |
