siRNA Details
| Gene Symbol: Wdr70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014750.1 | - | 4 | 305 | ||
| BC037184.1 | - | 4 | 309 | |||
| BC051482.1 | - | 4 | 275 | |||
| BC061236.1 | - | 4 | 308 | |||
| Gene Symbol: Gm1943 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK170003.1 | - | 1 | 294 | ||
| BC024407.1 | - | 1 | 287 | |||

Wdr70
Genomic Map