siRNA Details
| Gene Symbol: Mfsd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028657.3 | NP_082933.2 | 2 | 408 | ||
| GenBank mRNA | AK149678.1 | - | 2 | 414 | ||
| AK154386.1 | - | 2 | 410 | |||
| AK154902.1 | - | 2 | 488 | |||
| AK170185.1 | - | 2 | 428 | |||
| AK171084.1 | - | 2 | 410 | |||
| BC118620.1 | - | 2 | 408 | 40107417 | ||
| BC119790.1 | - | 2 | 408 | 40107416 | ||

Mfsd12
Genomic Map