siRNA Details
| Gene Symbol: Crtc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028881.2 | NP_083157.1 | 10, 11 | 1128 | ||
| GenBank mRNA | AK014553.1 | - | 5, 6 | 1499 | ||
| AK156192.1 | - | 10, 11 | 1107 | |||
| AK169944.1 | - | 6, 7 | 777 | |||
| BC023091.1 | - | 6, 7 | 718 | 4023064 | ||
| BC023831.1 | - | 10, 11 | 1123 | 5345301 | ||
| BC032183.1 | - | 10, 11 | 1133 | 5310152 | ||
| BC033300.1 | - | 10, 11 | 1123 | 4166411 | ||
| CV783724.1 | - | 7, 8 | 632 | |||

Crtc2
Genomic Map