siRNA Details
| Gene Symbol: A730049H05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043032.1 | - | 2, 3 | 371 | ||
| AK054516.1 | - | 1, 2 | 473 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A730049H05Rik| Gene Symbol: A730049H05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043032.1 | - | 2, 3 | 371 | ||
| AK054516.1 | - | 1, 2 | 473 | |||