siRNA Details
Gene Symbol: Tmem114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029070.2 | NP_083346.1 | 4 | 1059 | ||
GenBank mRNA | AK015757.1 | - | 4 | 1060 | ||
BC027071.1 | - | 4 | 1032 | |||
BC147190.1 | - | 4 | 836 | 9056234 | ||
BC147191.1 | - | 4 | 836 | 9056214 |
