siRNA Details
| Gene Symbol: Capsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029341.1 | NP_083617.2 | 4 | 555 | ||
| GenBank mRNA | AK006467.1 | - | 4 | 555 | ||
| BC037621.1 | - | 3 | 334 | |||
| BC061015.1 | - | 4 | 513 | 30297953 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Capsl| Gene Symbol: Capsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029341.1 | NP_083617.2 | 4 | 555 | ||
| GenBank mRNA | AK006467.1 | - | 4 | 555 | ||
| BC037621.1 | - | 3 | 334 | |||
| BC061015.1 | - | 4 | 513 | 30297953 | ||